Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p53 Ab-8 Mouse Monoclonal Antibody, Epredia™

Catalog No. p-4555631 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS738P1ABX 100 μL Concentrated 1mg/mL
MS738P1 500 μL Concentrated 200μg/mL
MS738P 1 mL Concentrated 200μg/mL
MS738PABX 200 μL Concentrated 1mg/mL
MS738R1 1 mL Prediluted
MS738R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS738P1ABX Supplier Epredia Supplier No. MS738P1ABX
Only null left
Add to Cart
Add to Cart

Deliver accurate, reproducible results with the Epredia™ p53 Ab-8, Mouse Monoclonal Antibody.

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: DO-7 + BP53-12

Isotype: IgG2b + IgG2a

Species Reactivity: Human. Others not known.

Epitope: aa 37-45 and aa 20-25

Immunogen: Recombinant human p53 protein

Molecular Weight: 53kDa

Positive Control: MDA-231, SKBR-3 cells or Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Specifications

Antigen p53 Ab-8
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DO-7 + BP53-12
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human p53 protein
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a, IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.