Learn More
CPC Scientific FITC-LC-Glu-Thr-Phe-Ser-Asp-Leu-Trp-Lys-Leu-Leu-NH2 (ammonium salt) 1MG
SDP
Supplier:  CPC Scientific CANC017A
Encompass
This is amino acids 17 to 26 fragment of p53, fluorescent labeled through an LC spacer. This peptide is the Mdm-2 binding domain of p53 known also as p53N. This sequence contains all of the residues that come into contact with the binding domain of Mdm-2. The tumor suppressor protein p53 is important in maintaining genome stability and in preventing cancer development, FITC (Abs/Em=493 nm/517 nm).. ONE-LETTER SEQUENCE: FITC-LC-ETFSDLWKLL-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:1753.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Yu, G. et al. PNAS 103, 1227 (2006); Wang, Y. et al. J. Biol. Chem. 281, 24803 (2006); Kanovsky, M. et al. PNAS 98, 12438 (2001)
Catalog No. 50-210-8894
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.