Learn More
Biogenex Laboratories p53 Protein : DO7 : 1mL
SDP
Supplier:  Biogenex Laboratories MU239UC
Encompass_Preferred
p53 is a tumor suppressor gene product identified in a wide variety of tumors. p53 protein is present in low concentration in normal cells, but elevated levels of mutantp53 have been found in many common cancers. Accumulation of mutant p53 detected by immunohistochemical staining has been reported in breast, lung, colon, stomach, bladder, and testis carcinomas, soft-tissue sarcomas, and melanomas.
Catalog No. 50-197-8880
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.