Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p53 (R175H Mutant) Recombinant Rabbit Monoclonal Antibody (HL1129)
GREENER_CHOICE

Catalog No. PIMA546935 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546935 Supplier Invitrogen™ Supplier No. MA546935
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

This gene encodes a tumor suppressor protein containing transcriptional activation, DNA binding, and oligomerization domains The encoded protein responds to diverse cellular stresses to regulate expression of target genes, thereby inducing cell cycle arrest, apoptosis, senescence, DNA repair, or changes in metabolism Mutations in this gene are associated with a variety of human cancers, including hereditary cancers such as Li-Fraumeni syndrome Alternative splicing of this gene and the use of alternate promoters result in multiple transcript variants and isoforms Additional isoforms have also been shown to result from the use of alternate translation initiation codons from identical transcript variants (PMIDs: 12032546, 20937277).
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53 (R175H Mutant)
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone HL1129
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Accession No. P29466, P48729, Q8N752
Gene Alias BCC7; BMFS5; LFS1; P53; TRP53; tumor protein p53
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide surrounding mutant R175H of human p53. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122011, 1452, 834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.