Learn More
Medchemexpress LLC HY-115576 10mg , P62-mediated mitophagy inducer CAS:1809031-84-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11557610MG
Encompass_Preferred
HY-115576 10mg P62-mediated mitophagy inducer CAS: 1809031-84-2 P62-mediated mitophagy inducer is a mitophagy regulator which activates mitophagy without recruiting Parkin or collapsing ΔΨm and retains activity in cells devoid of a fully functional PINK1/Parkin pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0573
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.