Learn More
MBL International Corporation Anti-p62 (SQSTM1) (Human) pAb (Polyclonal Antibody)
Supplier: MBL International Corporation PM045
Clonality:Polyclonal; Application:ICC|IHC|IP|WB; Conjugate:Unlabeled; Species Reactivity:Hamster|Human|Mouse|Rat; Size:100 µl; Target:p62; This antibody is validated for multiple applications (WB, IHC, ICC and IP) and is widely cited, including in research papers with more than 200 citations (Waguri S, Komatsu M. Biochemical and morphological detection of inclusion bodies in autophagy-deficient mice. Methods Enzymol. 453, 181-96 (2009)). This is a polyclonal antibody of 100 µl that is raised in rabbit and is reactive with human, hamster, mouse, rat.p62 Polyclonal Antibody.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.