Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ p70 S6 Kinase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580979
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580979 Supplier Invitrogen™ Supplier No. PA580979
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PS6K/RPS6KB1.

RPS6KB1 (p70S6K) is a serine/threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates several residues of the S6 ribosomal protein. p70 S6 kinase is predominantly localized in the cytoplasm, and is essential in growth factors regulated cell proliferation, pathways involving cell motility, such as metastases, the immune response, and tissue repair. p70 S6 kinase acts downstream of phosphoinositide (PI) 3-kinase, and its main physiological target is the S6 ribosomal protein, which is involved in upregulation of protein synthesis. The kinase activity of p70 S6 kinase leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding p70 S6 kinase and overexpression are seen in some breast cancer cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p70 S6 Kinase
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RPS6KB1
Gene Accession No. P23443
Gene Alias 10539; 2610318I15Rik; 4732464A07Rik; 70 kDa ribosomal protein S6 kinase 1; 7084; 70kDa; 7-10; AA959758; AI256796; AI314060; CG10539; CG10539-PA; CG10539-PB; Dmel\CG10539; Dmel_CG10539; Dp70[s6k]; Dp70S6k; dps6k; dS6 kinase; dS6K; d-S6K; dS6K1; EC 2.7.11.1; fs(3)07084; hypothetical protein; kinase p70S6K; KS6B1; l(3)07084; p70 p70-S6K; p70 ribosomal S6 kinase; p70 ribosomal S6 kinase alpha; p70 S6 kinase; p70 S6 kinase 1; p70 S6 kinase alpha; p70 S6 kinase, alpha; p70 S6K; p70 S6KA; p70 S6K-alpha; p70(S6K)-alpha; p70/85s6k; p70/S6K; p70[S6 kinase]; p70[S6k]; p70[S6kinase]; p70-alpha; p70S6 kinase; p70S6k; p70-S6K; p70-S6K 1; p70S6K kinase; p70s6k protein kinase homolog; P70S6K1; p70S6K-alpha; p80-S6K 1; p85 p85S6K; Pk64F; PkS6; protein kinase-like 64F; PS6K; p-S6K; ribosomal protein S6 kinase; ribosomal protein S6 kinase B1; ribosomal protein S6 kinase beta-1; Ribosomal protein S6 kinase I; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 70kD, polypeptide 1; ribosomal protein S6 kinase, 70kDa, polypeptide 1; ribosomal protein S6 kinase, polypeptide 1; ribosomal S6 protein kinase; Rps6kb1; RPS6-p70-protein kinase; S6 K; S6 kinase; s6k; S6K kinase; S6K1; s6k11; S6K-beta-1; S6kinase; S6-kinase; S6k-PA; S6k-PB; serine/threonine kinase 14 alpha; serine/threonine-protein kinase 14A; STK14A
Gene Symbols RPS6KB1
Host Species Rabbit
Immunogen Recombinant Human PS6K/RPS6KB1 Protein (Met1-Leu525).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6198
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.