Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p73 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501556948
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-6948 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-6948 Supplier Bethyl Laboratories, Inc Supplier No. A300126A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

p73 protein is a member of the p53 family of proteins. The tumor-suppressor protein p53 exhibits sequence specific DNA-binding, directly interacts with various cellular and viral proteins, and induces cell cycle arrest in response to DNA damage. In response to signals generated by a variety of genotoxic stresses, e.g, UV irradiation or DNA damage, p53 is expressed and undergoes post-translational modification that results in its accumulation in the nucleus. Activation of p53 leads to cell cycle arrest and in some cases to apoptosis, resulting in the inability of genetically damaged cells to proliferate. Thus, the p53-dependent pathways help to maintain genomic stability by eliminating damaged cells. The accumulation of high levels of p53 is a potential marker for maligcy.

Specifications

Antigen p73
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TP73
Gene Accession No. O15350
Gene Alias P73
Gene Symbols TP73
Host Species Rabbit
Immunogen Between 1 and 62
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7161
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.