Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Prazepam-d5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco P9051ML
Description
Prazepam is a benzodiazepine prescribed for the short-term treatment of anxiety. Sold under the trade name Centrax, this drug also has anticonvulsant, sedative, and muscle relaxant effects. This internal standard is applicable for quantitation of prazepam levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.
Specifications
| 67-56-1, 152477-89-9 | |
| MFCD00673307 | |
| 100 μg/mL in methanol | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
| C19D5H12ClN2O | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 329.83 | |
| 1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?Share a Content Correction