Learn More
Apexbio Technology LLC Ampicillin sodium(Synonyms: Ampicillin sodium salt, Sodium ampicillin, D-(-)-α-Aminobenzylpenicillin sodium salt), 10mM (in 1mL H2O), CAS: 69-52-3.
Supplier: Apexbio Technology LLC A25105.1
Ampicillin sodium (CAS 69-52-3) is a -lactam antibiotic that exerts antibacterial activity by competitively inhibiting bacterial transpeptidase enzymes Transpeptidases catalyze the critical final steps of bacterial cell wall biosynthesis their inhibition disrupts cell-wall integrity leading to bacterial cell lysis In assays using E coli 146 cells ampicillin sodium demonstrates an IC50 value of 1 8 g/ml against transpeptidase and an MIC value of 3 1 g/ml Research studies utilize ampicillin sodium to evaluate antibacterial efficacy in vitro and in animal infection models
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.