Learn More
Description
Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM (in both live and fixed cells) and STED.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 649 - 651 nm and 663 - 665 nm, respectively.
Please see the product protocol for further information and a guide to protein/antibody labeling.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Quantity | 300 μg |
| Product Type | Fluorescent Dye |
| Content And Storage | Store at -20°C |
| Target | Photoactivatable Dyes |
| Label Type | Other Labels or Dyes |
| Product Line | Tocris |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
