Learn More
PA2G4 Mouse anti-Human, Clone: OTI1D3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503203

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified.Specifications
PA2G4 | |
Monoclonal | |
Unconjugated | |
PA2G4 | |
EBP1, HG4-1, p38-2G4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5036 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1D3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UQ80 | |
PA2G4 | |
Full length human recombit protein of human PA2G4 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.