Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PABP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590094
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590094 Supplier Invitrogen™ Supplier No. PA590094
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTEMNGRIVA TKPLYVALAQ RKEERQAHLT NQYMQRMASV RAVPNPVINP YQPAPPSGYF MAAIPQTQNR AAYYPPSQIA QLRPSPRWTA QGARPHPFQN M; Positive Samples: A-549, HT-29, HeLa, Mouse brain, Rat lung; Cellular Location: Cytoplasm, Nucleus.

The poly(A)-binding protein (PABP), which is found complexed to the 3-prime poly(A) tail of eukaryotic mRNA, is required for poly(A) shortening and translation initiation. In humans, the PABPs comprise a small nuclear isoform and a conserved gene family that displays at least 3 functional proteins: PABP1 (PABPC1), inducible PABP (iPABP, or PABPC4), and PABP3 (PABPC3). In addition, there are at least 4 pseudogenes, PABPCP1 to PABPCP4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PABP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PABPC1
Gene Accession No. P11940, P29341, Q9EPH8
Gene Alias Cytoplasmic poly(A)-binding protein 1-A; ePAB; hypothetical protein; PAB1; pabp; PABP 1; Pabp1; PABP-1; PABP-1A; PABP-1-A; PABP-A; Pabpc1; pabpc1.L; pabpc1.S; pabpc1a; pabpc1-a; pabpc1a protein; pabpc1-b; pabpc2; PabpI; PABPL1; poly A binding protein 1; poly A binding protein, cytoplasmic 1 a; poly(A) binding protein cytoplasmic 1; poly(A) binding protein, cytoplasmic 1; poly(A) binding protein, cytoplasmic 1 L homeolog; poly(A) binding protein, cytoplasmic 1 S homeolog; poly(A) binding protein, cytoplasmic 1a; poly(A) binding protein, cytoplasmic 2; poly(A)-binding protein 1; poly(A)-binding protein 1A; poly(A)-binding protein 1-A; polyadenylate-binding protein 1; Polyadenylate-binding protein 1A; Polyadenylate-binding protein 1-A; polyadenylate-binding protein 1A; poly(A) binding protein, cytoplasmic 1a; wu:fb16a02; wu:fi19b08; wu:fj12d09; wu:fj61f06; XELAEV_18033915mg; xPABP1-A; zgc:109879
Gene Symbols PABPC1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human PABPC1 (NP_0025592).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171350, 18458, 26986
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.