Learn More
Selleck Chemical LLC PAC-1 10mg 315183-21-2
SDP
Supplier:  Selleck Chemical LLC S273810MG
Encompass_Preferred
PAC-1 is a potent procaspase-3 activator with EC50 of 0.22 ?M and the first small molecule known to directly activate procaspase-3 to caspase-3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2867