Learn More
CPC Scientific H-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-Gly-Lys-Arg-Tyr-Lys-Gln-Arg-Val-Lys-Asn-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PACA008B
Encompass
This peptide is the fragment 6-38 of PACAP, which is a much more potent and selective inhibitor of PACAP-27-stimulated pituitary adenylate cyclase than PACAP (6-27). Ki values for the inhibition of the enzyme were 7 nM and 150 nM respectively.ONE-LETTER SEQUENCE: FTDSYSRYRKQMAVKKYLAAVLGKRYKQRVKNK-NH2MOLECULAR FORMULA: C182H300N56O45S1MOLECULAR WEIGHT:4024.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [143748-18-9], SYNONYMS: Pituitary Adenylate Cyclase Activating Polypeptide-38 (6-38) (human, chicken, mouse, ovine, porcine, rat)RESEARCH AREA: HormonalREFERENCES: P. Robberecht et al., Mol. Pharmacol., 42, 347 (1992); P. Robberecht et al., Eur. J. Biochem., 207, 239 (1992); F. Ohno et al., Regul. Peptides, 126, 115 (2005); P. Robberecht, P. Gourlet, P. De Neef, M-C. Woussen-Colle, M-C. Vandermeers-Piret,A. Vandermeers, and J. Christophe, Eur. J. Biochem., 207, 239 (1992) (Original); A. Vandermeers, S. Vandenborre, X. Hou, P. De Neef, P. Robberecht, M-C. Vandermeers-Piret,an
Catalog No. 50-211-0067