Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PACAP-38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143860 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143860 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143860 Supplier Invitrogen™ Supplier No. PA5143860
  • $470.00
    $415.65 / Each of 1

    Save $54.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEL whole cell. IHC: human lung cancer tissue. Flow: HEL cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Binding to its receptor activates G proteins and stimulates adenylate cyclase in pituitary cells. Promotes neuron projection development through the RAPGEF2/Rap1/B-Raf/ERK pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACAP-38
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Adcyap1
Gene Accession No. P18509
Gene Alias Adcyap1; adenylate cyclase activating polypeptide 1; adenylate cyclase activating polypeptide 1 (pituitary); PACAP; PACAP-27; PACAP-38; PACAP-related peptide; pituitary adenylate cyclase activating polypeptide; pituitary adenylate cyclase activating polypeptide 1; pituitary adenylate cyclase-activating polypeptide; Pituitary adenylate cyclase-activating polypeptide 27; Pituitary adenylate cyclase-activating polypeptide 38; pituitary adenylate cyclase-activating polypeptide precursor (Pacap); prepro-PACAP; PRP-48
Gene Symbols Adcyap1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human PACAP-38/ADCYAP1 (QMAVKKYLAAVLGKRYKQRVK).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.