Learn More
CPC Scientific H-His-Ser-Asp-Gly-Ile-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-Gly-Lys-Arg-Tyr-Lys-Gln-Arg-Val-Lys-Asn-Lys-NH2 1MG
SDP
Supplier:  CPC Scientific PACA002B
Encompass
SEQUENCE: H-His-Ser-Asp-Gly-Ile-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-Gly-Lys-Arg-Tyr-Lys-Gln-Arg-Val-Lys-Asn-Lys-NH2ONE-LETTER SEQUENCE: HSDGIFTDSYSRYRKQMAVKKYLAAVLGKRYKQRVKNK-NH2MOLECULAR FORMULA: C203H331N63O53S1MOLECULAR WEIGHT: 4534.32STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [124123-15-5]SYNONYMS: Pituitary Adenylate Cyclase Activating Polypeptide-38 (human, mouse, ovine, porcine, rat), PACAP-38 (human, mouse, ovine, porcine, rat)RESEARCH AREA: HormonalREFERENCES:A. Miyata et al., BBRC, 164, 567 (1989)K. Ogi et al., BBRC, 173, 1271 (1990)SKU(s): PACA-002A, PACA-002B
Catalog No. 50-211-0055