Learn More
CPC Scientific H-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PACA005A
Encompass
A specific inhibitor of adenylate cyclase activation that results in clear functional discrimination between PACAP-27 receptors and PACAP-38 receptors.SEQUENCE: H-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: FTDSYSRYRKQMAVKKYLAAVL-NH2MOLECULAR FORMULA: C121H193N33O31S1MOLECULAR WEIGHT: 2638.2STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [136134-68-4]SYNONYMS: RESEARCH AREA: HormonalREFERENCES:P. Robberecht et al., FEBS Letters, 286, 133 (1991)SKU(s): PACA-005A, PACA-005B
Catalog No. 50-211-0060
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.