Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PACAP Monoclonal Antibody (1996CT329.45.62)
GREENER_CHOICE

Catalog No. PIMA537769 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537769 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537769 Supplier Invitrogen™ Supplier No. MA537769
  • $599.00
    $498.65 / Each of 1

    Save $100.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

PACAP Monoclonal Antibody for Western Blot

ADCYAP1 is adenylate cyclase activating polypeptide 1. Mediated by adenylate cyclase activating polypeptide 1 receptors, this polypeptide stimulates adenylate cyclase and subsequently increases the cAMP level in target cells. Adenylate cyclase activating polypeptide 1 is not only a hypophysiotropic hormone, but also functions as a neurotransmitter and neuromodulator. In addition, it plays a role in paracrine and autocrine regulation of certain types of cells. This gene encodes three different mature peptides, including two isotypes, a shorter form and a longer form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACAP
Applications Western Blot
Classification Monoclonal
Clone 1996CT329.45.62
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MZB1
Gene Accession No. Q8WU39
Gene Alias ADCYAP1; adenylate cyclase activating polypeptide 1; adenylate cyclase activating polypeptide 1 (pituitary); HSPC190; MGC126852; MZB1; OTTHUMP00000162201; PACAP; pacap 38; PACAP-27; PACAP-38; PACAP-related peptide; pERp1; pituitary adenylate cyclase activating polypeptide; pituitary adenylate cyclase activating polypeptide 1; pituitary adenylate cyclase-activating polypeptide; Pituitary adenylate cyclase-activating polypeptide 27; Pituitary adenylate cyclase-activating polypeptide 38; pituitary adenylate cyclase-activating polypeptide precursor (Pacap); prepro-PACAP; PRP-48
Gene Symbols MZB1
Host Species Mouse
Immunogen recombinant protein from the human region of human ADCYAP1.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51237
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.