Learn More
CPC Scientific H-Asp-Val-Ala-His-Glu-Ile-Leu-Asn-Glu-Ala-Tyr-Arg-Lys-Val-Leu-Asp-Gln-Leu-Ser-Ala-Arg-Lys-Tyr-Leu-Gln-Ser-Met-Val-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PACA006B
Encompass
Hypothalamic regulatory peptide.ONE-LETTER SEQUENCE: DVAHEILNEAYRKVLDQLSARKYLQSMVAMOLECULAR FORMULA: C148H242N42O45S1MOLECULAR WEIGHT:3361.87STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [132769-35-8], SYNONYMS: PACAP Related Peptide (1-29) (rat)RESEARCH AREA: HormonalREFERENCES: K. Ogi et al., BBRC, 173, 1271 (1990)
Catalog No. 50-211-0063
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.