Learn More
Biolegend Pacific Blue™ anti-mouse CD38 [S21016F]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Supplier:  Biolegend 165615
Encompass_Preferred
Pacific Blue™ anti-mouse CD38 [S21016F]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-272-3393
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.