Learn More
Thermo Scientific™ Pacific Hemostasis™ Factor Assays
Catalog No. 22176461 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Factor VIII Deficient Plasma
Catalog No. Type
22-176461 Factor VIII Deficient Plasma
1 options
This item is not returnable. View return policy
Catalog No. 22-176461
This item is not returnable. View return policy
Fisherbrand™ Pacific Hemostasis Factor Assays are used for the detection and quantification of factor deficiencies in patient plasmas.
Detect and quantify factor deficiencies in patient plasmas with Thermo Scientific™ Pacific Hemostasis™ Factor Assays.
Pacific Hemostasis Factor Assays include a complete line of lyophilized congenital Factor Deficient Plasmas for performing factor assays.
  • All factor deficient plasmas are prepared from congenitally deficient patient donors or immunodepleted plasma, with the exception of Factor II
  • Factor II Deficient Plasma is a mix of human and bovine plasma
  • Plasmas are packaged 10 × 1mL; Diluting Fluid is packaged 2 × 100mL
TRUSTED_SUSTAINABILITY
Specifications
Type Factor VIII Deficient Plasma