Learn More
Thermo Scientific™ Pacific Hemostasis™ FDP Assay
Catalog No. 23249989
Change view
Click to view available options
Quantity:
2 x 100 mL
30 Determinations
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-249989 30 Determinations
23-262567 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-249989
This item is not returnable. View return policy
Fisherbrand™ Pacific Hemostasis FDP Assay is a fast, easy-to-use latex agglutination assay for use in measuring fibrinogen degradation products (FDP) in serum and urine.
Measure fibrinogen degradation products (FDP) in serum and urine with Thermo Scientific™ Pacific Hemostasis™ FDP Assay, a fast, easy-to-use latex agglutination assay used in the diagnosis of disseminated intravascular coagulation (DIC), deep vein thrombosis (DVT), pulmonary embolism (PE), certain pregnancy disorders and myocardial infarction.
Features:
  • Latex sensitivity of 1ug/mL of FDP
  • No pretreatment of heparin specimen
  • No repeat testing due to false positives from heparin interference
TRUSTED_SUSTAINABILITY
Specifications
Description Assay Kit
Quantity 30 Determinations