Learn More
Eisco Labs Pack of 12 Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
Shop All Eisco Products

Description
- Pack of 12 reagent bottles: Includes 12 wide-mouth amber HDPE bottles with polypropylene caps, designed for lab storage and secure transport of light-sensitive samples.
- Light-shielding design: Amber color protects light-sensitive substances by blocking UV rays, ensuring safe storage of photosensitive materials in labs.
- Leak-proof construction: Bottles are leak-tested, offering secure containment for various chemicals and reagents.
- Chemical-resistant materials: Constructed from durable HDPE, these bottles offer excellent chemical resistance for versatile lab and industrial applications.
- Convenient and reliable: Wide neck allows easy filling and pouring.
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Amber |
| Diameter (Metric) Inner Neck | 33.1 mm |
| Material | HDPE |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Height (Metric) | 130 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.