Learn More
Teledyne ISCO, Inc Pack-In-The-Box Kit: Cp Class Dual Piston Pump With Belt Drive And Shaped Cams; 0.00 - 12.00 Ml/Min.
Supplier:  Teledyne ISCO, Inc CP012BRX
Encompass
Pack-In-The-Box Kit: CP Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.00 - 12.00 mL/min.; 18,000 psi; Stainless Steel Fluid Path; Automatic Piston Wash; with Pressure & Leak Sensors; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9933
Only null left
Add to Cart
Request a Quote