Learn More
Thermo Scientific™ Cell Locker™ Chamber with Copper Tray and Transport Cover
Catalog No. 11675975
Change view
Click to view available options
No. of Places:
6
Catalog No. No. of Places
11-675-975 6
1 options
Catalog No. 11-675-975 Supplier Thermo Scientific™ Supplier No. 50154739X6
Add to Cart
Add to Cart
Request a Quote
Protect your most sensitive cells, even during transport.
Each individual Cell Locker™ Chamber is effectively a quarantine chamber. Can be manually disinfected or can be autoclaved.
  • Dual 0.2 μm membrane filters permit air circulation but exclude microbial contaminants
  • Replacable 0.2 μm membrane filter is hydrophobic, oleophobic, resistant to organic solvents and tested for biosafety and low cytotoxicity
  • Dishes and flasks can be removed individually on a work tray or inside the entire chamber with optional transport cover, thus protecting samples from outside air
  • Non-skid tray slides out to allow access to culture vessels, or the entire tray can be removed
TRUSTED_SUSTAINABILITY
Specifications
No. of Places 6
For Use With (Application) Cell Protection and Transport
Includes Chamber, Copper Tray, and Transport Cover
Quantity 1 Ea.