Learn More
MilliporeSigma™ Supelco™ Packed Column Ferrule Kit
Catalog No. 111011025 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packed Column Ferrule Kit
Catalog No.
11-101-1025 Packed Column Ferrule Kit
1 options
Catalog No. 11-101-1025 Supplier MilliporeSigma™ Supelco™ Supplier No. 20648
Ferrule Test Kit for 1/4 in. O.D. Glass Packed Columns: 4 Supeltex M-1, 4 Supeltex M-2A, 4 Supeltex M-4 ferrules, a 6 in./15 cm x 1/4 in. practice piece of glass tubing, and instructions.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Chromatography Applications