Learn More
Selleck Chemical LLC Paclitaxel 50mg 33069-62-4 NSC 125973, PTX, Taxol, Onxal
SDP
Supplier:  Selleck Chemical LLC S115050MG
Encompass_Preferred
Paclitaxel (NSC 125973, PTX, Taxol, Onxal) is a microtubule polymer stabilizer with IC50 of 0.1 pM in human endothelial cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0918319
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.