Learn More
Hach Company Pad, absorbent, w/ dispenser (10 pks of 100)
Supplier: Hach Company 1491800
Dispense a clean cellulose pad into a petri dish quickly and easily with this absorbent pad kit. 100, sterile, 47 mm pads are packed in each tube. Each tube slips quickly into the provided hand dispenser for easy use. Gamma radiation sterilization makes pads suitable for exacting microbiological procedures and eliminates hazardous ETO residuals. One hand dispensing allows transfer of pad to petri dish without handling. Pall Life Sciences