Learn More
Med Vet International BD Vacutainer Venous Blood Collection Tube, Sodium Citrate Additive, 1.8 mL, Hemogard™ Closure, Polyethylene Terephthalate (PET) Tube, 100/Box
Supplier:  Med Vet International 363080
Encompass_Preferred
Features Consists of PET (Polyethylene Terephthalate), the virtually unbreakable material used for all BD Plastic Tubes (outer tube) Retains vacuum, maintaining proper blood to additive ratio Clear BD Hemogard™ closure for 1.8mL tubes BD Vacutainer® Citrate Tubes with 3.2% buffered sodium citrate solution are used for routine coagulation studie
Catalog No. 50-304-5000
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.