Learn More
Essendant PADCLEANER13BE
Supplier:  Essendant MMM08406
Encompass_Preferred
3M Blue Cleaner Pad 5300 is designed to remove worn finish soils scuffs and spills and to prepare surfaces for recoating The pad is constructed of high quality nylon and polyester synthetic fibers in a non-woven web A durable resin coating system permeates the fibers throughout the pad with abrasive cleaning particles providing consistent performance over the life of the pad The resin coating will not be significantly affected by water detergents and cleaners normally used for floor maintenance The floor scrubbing pad is reversible Depending on the surface and application the floor pad can be used with detergent solutions for dry scrubbing floors and in heavy duty spray cleaning operations
Catalog No. 50-000-33348
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.