Learn More
Essendant PADDIAMONDTZABRASIVRD
Supplier:  Essendant MMM86026
Encompass_Preferred
Diamond disc system for more aggressive refurbishing of stone floors with apparent wear or damage from scratching or pitting caused by ice melting chemicals or wear from pallets heavy traffic or other mechanical damage Creates a smooth highly refined surface ready for finishing with Scotchgard Stone Floor Protector Consistently and rapidly smoothes and levels stone floor surfaces For use on low speed rotary or automatic equipment
Catalog No. 50-000-31440
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.