Learn More
3M™ Paint Masking Tape
Catalog No. 19072089 Shop All 3M Products
Change view
Click to view available options
Quantity:
24/Cs.
Catalog No. Quantity
19-072-089 24/Cs.
1 options
Catalog No. 19-072-089 Supplier 3M™ Supplier No. 7000088515
Add to Cart
Add to Cart
3M™ Paint Masking Tape is a premium high-performance crepe paper masking tape designed to meet aerospace needs in most painting applications, including the use of multiple bake cycles.
  • Rubber adhesive provides strong tack and adhesion, and resists lifting
  • Clean removal from many surfaces following multi-bake or high temperature bake
  • Smooth mini-crepe paper backing conforms and maintains integrity when formed around a corner
  • Strong backing is easy to tear, yet resists slivering and provides one-piece removal
  • Controlled unwind dispenses evenly, reducing fatigue
TRUSTED_SUSTAINABILITY
Specifications
Material Rubber
Product Type Masking Tape
For Use With (Application) Painting and Depainting
Model 231
Quantity 24/Cs.
Thickness (English) 7.6 mil
Dimensions (L x W) 55 x 36 mm