Learn More
Fisherbrand™ Painted Steel Vacuum Pump Cart
Catalog No. 15050072 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
16 x 19 x 5.7 in.
16 x 31 x 5.7 in.
16 x 43 x 5.7 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
15-050-072 16 x 31 x 5.7 in.
15-050-071 16 x 19 x 5.7 in.
15-050-073 16 x 43 x 5.7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 15-050-072
Add to Cart
Add to Cart
This item is not returnable. View return policy
Vacuum Pump Cabinets are standing height, 35.1 in. tall. They have sound deadening material included. Pump cart is sold separately. Standard color is Fisherbrand™ White.
  • Vacuum Pump Cart
  • Fisherbrand White is a urethane powder coating that provides reliable chemical and abrasion resistance
  • Locking swivel casters for ease of use to access vacuum pumps within cabinets
Specifications
Color White
Dimensions (L x W x H) 16 x 31 x 5.7 in.
Material Steel (18 ga.)