Learn More
United Scientific™ Pair of Photogates with Brackets
SDPSCI_ED
Catalog No. S139509 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Equipment):
Air Track
Catalog No. For Use With (Equipment)
S139509 Air Track
1 options
This item is not returnable. View return policy
Catalog No. S139509 Supplier United Scientific™ Supplier No. DTPHG1P
Add to Cart
Add to Cart
This item is not returnable. View return policy
Photogates that can be used with free fall apparatuses, air tracks, and much more.
Designed to be used with air tracks, free-fall apparatus and other equipment.
  • Offers eight programmed functions to record counts, time intervals, periods of oscillation, accelerations, acceleration due to gravity and velocities in collisions
  • Comes with two photogates
Specifications
For Use With (Equipment) Air Track
Product Type Photogates
Prop 65 Warning: Cancer and reproductive harm