Learn More
Carnabio Usa Inc PAK5(PAK7)/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 071275
Encompass_Preferred
PAK5(PAK7), Catalytic domain, Wild type, Amino Acid 425-719, NP_065074.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0557
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.