Learn More
Leap PAL Parts Pal 40Ml Clear Glass Vial 24Mm
Supplier:  Leap PAL Parts VIAL40ND24CG100
Encompass_Preferred
PAL System Vial 40CV 40ml Clear Glass with Label designed for the PAL Autosampler. 95x27.5mm 1st Class Hydrolytic Glass with flat top for optimized sealing. For ND24 Screw Caps Pk of 100 Pcs
Catalog No. 501920904
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.