Learn More
Bachem Palmitoyl-Gly-His-Lys-OH
SDP
Supplier:  Bachem 4142828.2005
Encompass_Preferred
Pal-GHK or Palmitoyl Tripeptide-1 is a version of GHK peptide (copper-binding peptide) designed for better skin penetration, since it is a messenger peptide for collagen renewal. Therefore Pal-GHK leads to wrinkle reduction, skin firming and skin improvement.
Catalog No. NC2404498
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.