Learn More
Sigma Organic Chemistry Palladium On Alumina Catalytic Static Mixer 316L 6mm Dia X 150mm | 1Ea
Supplier: Sigma Organic Chemistry 9197131EA

PALLADIUM ON ALUMINA CATALYTIC STATIC MIXER 316L 6MM DIA X 150MM, 1EA
About This Item:
UNSPSC Code: 12352200
Storage: ROOM TEMPERATURE
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.