Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palatine Chrome Green, TCI America™
SDP

Catalog No. P059425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P059425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P059425G Supplier TCI America Supplier No. P059425G

Chemical Identifiers

CAS 3564-28-1
Molecular Formula C16H12N4Na2O10S2
Molecular Weight (g/mol) 530.39
MDL Number MFCD00135102
InChI Key SIOTWGZRRGSLRA-AXHWYJOWSA-N
Synonym Mordant Green 17
PubChem CID 131852861
IUPAC Name (3E)-5-amino-3-[(2-hydroxy-5-nitrophenyl)hydrazinylidene]-4-oxonaphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC(=C(C=C1[N+](=O)[O-])NN=C2C(=CC3=CC(=CC(=C3C2=O)N)S(=O)(=O)O)S(=O)(=O)O)O.[Na].[Na]

Specifications

Color Black
Quantity 25 g
Formula Weight 528.37
Physical Form Crystalline Powder
Chemical Name or Material Palatine Chrome Green
EINECSNumber (5)-2097
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.