Learn More
Palatinose hydrate, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 343336-76-5 |
| Linear Formula | C12H22O11 · xH2 O |
| MDL Number | MFCD11100373 |
| Synonym | 6-O-alpha-D-Glucopyranosyl-D-fructose; Isomaltulose hydrate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 342.30 (anhydrous basis) |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.