Learn More
Medchemexpress LLC HY-50767A 500mg , Palbociclib (hydrochloride) PD 0332991 hydrochloride CAS:827022-32-2 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY50767A500MG
Encompass_Preferred
Medchemexpress, HY-50767A 500mg Palbociclib (hydrochloride) PD 0332991 hydrochloride CAS:827022-32-2 Formula:C24H30ClN7O2 IC50: 11 nM (Cdk4), 16 nM (Cdk6) Purity:98% Palbociclib hydrochloride is a highly specific inhibitor of Cdk4 ( IC 50 =11 nM) and Cdk6 ( IC 50 =16 nM), having no activity against a panel of 36 additional protein kinases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2322
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.