Learn More
Medchemexpress LLC HY-A0019A 10mg Medchemexpress, Paliperidone palmitate CAS:199739-10-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0019A10MG
Encompass_Preferred
Medchemexpress, HY-A0019A 10mg Paliperidone palmitate CAS:199739-10-1 Paliperidone palmitate (9-Hydroxyrisperidone palmitate), an atypical long-acting antipsychotic agent, is an ester prodrug of Paliperidone. Paliperidone is a dopamine antagonist and 5-HT2A antagonist of the atypical antipsychotic class. Paliperidone palmitate shows efficacy against schizophrenia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0997
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.