Learn More
Cytiva HEPA Capsule with Versapor Membrane - 1.2 µm, 3/8 in. FNPT & hose barb (1/pkg)
Supplier:  Cytiva 12144
Encompass_Preferred
Ensures High Efficiency Particulate-Free Air (HEPA)-Porous membrane delivers high air flow rates at low differential pressure and extends filter life-Ensures particulate-free air 99.97% retention of 0.3 µm DOP aerosol following ASTM D 2986-95A This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0501487
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.