Learn More
Invitrogen™ Palladin Monoclonal Antibody (1E6)

Description
Suggested positive control: MDA-MB231 (human breast cancer) lysates.
Specifications
Specifications
| Antigen | Palladin |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1E6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% sodium azide |
| Gene | PALLD |
| Gene Accession No. | P0C5E3, Q8WX93, Q9ET54 |
| Gene Alias | 2410003B16Rik; 6030492A02; 90kDa actin-associated protein palladin; actin-associated protein palladin; CGI151; CGI-151; FLJ22190; FLJ38193; FLJ39139; involved in cytoskeletal assembly; KIAA0992; MYN; myoneurin; palladin; palladin isoform; palladin pseudogene; palladin, cytoskeletal associated protein; PALLD; PNCA1; Sarcoma antigen NY-SAR-77; SIH002 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.