Learn More
Palladium, 5% on calcium carbonate, Type A306060-5, lead poisoned
Description
Selective hydrogenationPalladium (5%) on calcium carbonate powder (reduced) is a heterogeneous and a mild catalyst, used in the hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic carbonyls, olefins, and nitriles. It has high selectivity towards certain chemical reactants, and is often the preferred heterogeneous catalyst for a wide variety of chemical reactions. Partially deactivated form of palladiuim (5%) on calcium carbonate is known as Lindlar catalyst, which is useful for chemo and stereoselective hydrogenations, such as alkynes to cis-alkenes (for e.g., Vitamin synthesis).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 5 g |
| UN Number | UN3077 |
| Solubility Information | Insoluble in most organic solvents. |
| Chemical Name or Material | Palladium, 5% on calcium carbonate, Type A306060-5, Lead poisoned |
Safety and Handling
| Hazard Category | H350-H360Df |
| Hazard Statement | GHS H Statement H360-H201 May damage fertility or the unborn child. Explosive mass explosion hazard. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.