Learn More
Sigma Organic Chemistry Palladium on alumina | 50G | MFCD03613602
Supplier: Sigma Organic Chemistry 76117650G

Palladium on alumina, 50G
About This Item:
Linear Formula: Pd
MDL Number: MFCD03613602
UNSPSC Code: 12352300
Storage: room temp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.