Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) chloride, 99.999%, (trace metal basis), Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
7647-10-1
Cl2Pd
177.31
PIBWKRNGBLPSSY-UHFFFAOYSA-L
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
24290
CHEBI:53434
dichloropalladium
Cl[Pd]Cl
7647-10-1
Cl2Pd
177.31
PIBWKRNGBLPSSY-UHFFFAOYSA-L
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
24290
CHEBI:53434
dichloropalladium
Cl[Pd]Cl
For ultra trace analysis
Assay Percent Range 59 to 61% (Pd)
Linear Formula PdCl2
Merck Index 15,7091
Solubility Information Solubility in water: 31.99g/L. Other solubilities: soluble in hydrochloric acid,hydrobromic acid,soluble in alcohol and acetone
Formula Weight 177.31
Physical Form Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid, hydrobromic acid, soluble in alcohol and acetone
Total Trace Metal Impurities 0.001% max
Color White
Melting Point 678.0°C to 680.0°C
Quantity 1g
Chemical Name or Material Palladium(II) chloride
Show More Show Less

GHS H Statement
May be corrosive to metals.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 231-596-2