Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) chloride, solution, Pd 9.0-11.0% w/w (cont. Pd)

Catalog No. p-7049059
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12623NH 1 g
AA12623NB 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA12623NH Supplier Thermo Scientific Chemicals Supplier No. 012623NH
Only null left
Add to Cart

CAS: 7647-10-1 | Cl2Pd | 177.32 g/mol

Palladium(II) chloride is used to remove the stain present stainless steel . Its solutions are used to test the corrosion-resistance of stainless steel. It is used in carbon monoxide detectors. It is also used for cosmetic tatooing of leukomas in thecornea and in the preparation of palladium(II) iodide by reacting with potassium iodide. It is involved in the synthesis of semiconducting metal-containing polymers. It is used as a catalyst for carbonylation of organic tellurides and deamination of phenethylamines to phenyl substituted pyrroles with copper(I) as co catalyst. It plays an important role in the hydrodimerization of butadiene to form octa-2,7-dien-1-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-10-1
Molecular Formula Cl2Pd
Molecular Weight (g/mol) 177.32
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
Synonym palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PubChem CID 24290
ChEBI CHEBI:53434
IUPAC Name dichloropalladium
SMILES [Cl-].[Cl-].[Pd++]

Specifications

Color Brown
UN Number UN3264
Concentration Pd 9.0 to 11.0% w/w (cont. Pd)
Solubility Information Miscible with water,organic solvents and hydrochloric acid.
Formula Weight 177.31
Odor Acidic
Quantity 1 g
Physical Form Liquid
Chemical Name or Material Palladium(II) chloride, Solution
Hazard Category H290-H317-H318
Hazard Statement GHS H Statement
H314-H318-H290-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 231-596-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.