Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palm Oil, Spectrum™ Chemical
SDP

Catalog No. 18604945 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
25 kg
200 L
Packaging:
Poly Bottle
Poly Drum
Steel Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-945 2.5 kg Poly Bottle
18-604-946 200 L Steel Drum
18-604-947 25 kg Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-945 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PA1052.5KG
Add to Cart
Add to Cart

PA105, 8002-75-3

Spectrum™ Chemical Palm Oil is a vegetable oil that is edible and comes from the mesocarp of the oil palms. It can also be used to make biodiesel. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 8002-75-3

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 2.5 kg
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.